Guaiacol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, 2-Methoxyphenol
M-Clarity™ quality level = MQ100
G5502-100G
G5502-1KG
G5502-250G
SIALG5502-100GEA
49.6
EUR
SIALG5502-100G
SIALG5502-1KG
SIALG5502-250G
Guaiacol, Sigma-Aldrich®
Guaiacol
Formula:
C₇H₈O₂ Boiling Pt: 205 °C (1013 hPa) Melting Pt: 28…32 °C Density: 1.1534 g/cm³ (0 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002185 CAS Number: 90-05-1 EINECS: 201-964-7 UN: 2810 ADR: 6.1,III Merck Index: 13,04568 |