2,4-Dibromopyridine, Sigma-Aldrich®

Supplier: Merck
Warning

725196-1G 725196-5G
SIAL725196-1GEA 29.2 EUR
SIAL725196-1G SIAL725196-5G
2,4-Dibromopyridine, Sigma-Aldrich®
2,4-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
MDL Number: MFCD01859720
CAS Number: 58530-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR