Manganese(II) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Manganese dichloride
M-Clarity™ quality level = MQ200
244589-10G 244589-250G 244589-50G
SIAL244589-10GEA 70.5 EUR
SIAL244589-10G SIAL244589-250G SIAL244589-50G
Manganese(II) chloride, Sigma-Aldrich®
Manganese (II) chloride
Formula: MnCl₂
MW: 125.84 g/mol
Boiling Pt: 1190
Melting Pt: 650
Density: 2.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1 III
Merck Index: 12,05768