Oleoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
367850-100ML 367850-500ML
SIAL367850-100MLEA 80.1 EUR
SIAL367850-100ML SIAL367850-500ML
Oleoyl chloride, Sigma-Aldrich®
Oleoyl chloride
Formula: C₁₈H₃₃ClO
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.9147 g/cm³ (20 °C)
Flash Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00134332
CAS Number: 112-77-6
EINECS: 204-005-0
UN: 3265
ADR: 8,II