Bis(2-ethylhexyl)amine, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dioctylamine, Di(2-ethylhexyl)amine, Bis(2-ethylhexyl)amine
M-Clarity™ quality level = MQ100
292850-1L 292850-250ML
SIAL292850-1LEA 146 EUR
SIAL292850-1L SIAL292850-250ML
Bis(2-ethylhexyl)amine, Sigma-Aldrich®
Bis(2-ethylhexyl) amine
Formula: C₁₆H₃₅N
MW: 241.46 g/mol
Boiling Pt: 281.1 °C (1013 hPa)
Melting Pt: < –60 °C
Density: 0.8062 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00009489
CAS Number: 106-20-7
EINECS: 203-372-4
UN: 2922
ADR: 8,II