(±)-3-Buten-2-ol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: (±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol

M-Clarity™ quality level = MQ100

B86400-25G B86400-5G
SIALB86400-25GEA 149 EUR
SIALB86400-25G SIALB86400-5G
(±)-3-Buten-2-ol, Sigma-Aldrich®
(±)-3-Buten-2-ol
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 96
Melting Pt: –100 °C
Density: 0.913 g/cm³ (20 °C)
Flash Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00004543
CAS Number: 598-32-3
EINECS: 209-929-8
UN: 1986
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR