Benzenesulphonic acid
Supplier: Merck
Danger
93792-500G 93792-1KG
SIAL93792-500GEA 1970 EUR
SIAL93792-500G SIAL93792-1KG
Benzenesulphonic acid
Benzenesulphonic acid
Formula: C₆H₆O₃S
MW: 158.18 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 50 °C
Density: 1.32 g/cm³ (47 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011689
CAS Number: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8,III
Merck Index: 13,01070