3-Methyl-3-buten-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Methylbut-3-en-1-ol, 2-Methyl-1-buten-4-ol
M-Clarity™ quality level = MQ300
W519308-4KG
W519308-SAMPLE
SIALW519308-4KGEA
414
EUR
SIALW519308-4KG
SIALW519308-SAMPLE
3-Methyl-3-buten-1-ol, Sigma-Aldrich®
3-Methyl-3-buten-1-ol
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 132 °C (1013 hPa) Melting Pt: –82 °C Density: 0.853 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002933 CAS Number: 763-32-6 EINECS: 212-110-8 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...