2,3-Difluoropyridine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
718173-1G
718173-5G
SIAL718173-1GEA
26.1
EUR
SIAL718173-1G
SIAL718173-5G
2,3-Difluoropyridine, Sigma-Aldrich®
2,3-Difluoropyridine
Formula:
C₅H₃F₂N MW: 115.08 g/mol |
MDL Number:
MFCD04114143 CAS Number: 1513-66-2 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...