3-Isobutyl-1-methylxanthine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,6-Dihydroxy-3-isobutyl-1-methylpurine, IBMX, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione
I7018-100MG I7018-250MG I7018-1G
SIALI7018-100MGEA 193 EUR
SIALI7018-100MG SIALI7018-250MG SIALI7018-1G
3-Isobutyl-1-methylxanthine, Sigma-Aldrich®
3-Isobutyl-1-methylxanthine
Formula: C₁₀H₁₄N₄O₂
MW: 222.25 g/mol
Melting Pt: 200…201 °C
MDL Number: MFCD00005584
CAS Number: 28822-58-4
EINECS: 249-259-3