2-Amino-6-fluoropyridine, Sigma-Aldrich®

Supplier: Merck
Warning

725064-1G 725064-5G
SIAL725064-1GEA 40.5 EUR
SIAL725064-1G SIAL725064-5G
2-Amino-6-fluoropyridine, Sigma-Aldrich®
2-Amino-6-fluoropyridine
Formula: C₅H₅FN₂
MW: 112.11 g/mol
MDL Number: MFCD04114200
CAS Number: 1597-32-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR