2-Methyl-2-butanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: tert-Amyl alcohol, tert-Pentyl alcohol, tert-Pentanol
721123-100ML 721123-1L
SIAL721123-100MLEA 97.5 EUR
SIAL721123-100ML SIAL721123-1L
2-Methyl-2-butanol, Sigma-Aldrich®
2-Methyl-2-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –8.4 °C
Density: 0.806 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00004478
CAS Number: 75-85-4
EINECS: 200-908-9
UN: 1105
ADR: 3,II
Merck Index: 13,07218