4-Iodobenzoyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: p-Iodobenzoyl chloride

M-Clarity™ quality level = MQ100

252123-25G 252123-5G
SIAL252123-25GEA 102 EUR
SIAL252123-25G SIAL252123-5G
4-Iodobenzoyl chloride, Sigma-Aldrich®
4-Iodobenzoyl chloride
Formula: C₇H₄ClIO
Boiling Pt: 100 °C (0.7 mmHg)
Melting Pt: 63…65 °C
MDL Number: MFCD00001058
CAS Number: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR