4-Pentyn-1-ol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Pentyn-5-ol
M-Clarity™ quality level = MQ100
302481-25G
302481-5G
SIAL302481-25GEA
191
EUR
SIAL302481-25G
SIAL302481-5G
4-Pentyn-1-ol, Sigma-Aldrich®
4-Pentyn-1-ol
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 50 °C (1013 hPa) Density: 0.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002974 CAS Number: 5390-04-5 EINECS: 226-383-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...