Propionic anhydride, Sigma-Aldrich®
Supplier: Merck
Danger
P51478-1KG P51478-500G
SIALP51478-1KGEA 85.7 EUR
SIALP51478-1KG SIALP51478-500G
Propionic anhydride, Sigma-Aldrich®
Propionic anhydride
Formula: C₆H₁₀O₃
Boiling Pt: 167 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.01 g/cm³ (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00009303
CAS Number: 123-62-6
EINECS: 204-638-2
UN: 2496
ADR: 8,III
Merck Index: 12,08011