1,4-Butane diisothiocyanate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,4-Butylene diisothiocyanate
708909-1G 708909-5G
SIAL708909-1GEA 76.8 EUR
SIAL708909-1G SIAL708909-5G
1,4-Butane diisothiocyanate, Sigma-Aldrich®
1,4-Butane diisothiocyanate
Formula: C₆H₈N₂S₂
MW: 172.27 g/mol
Boiling Pt: 102…104 °C (0.1 mmHg)
Melting Pt: 124 °C
MDL Number: MFCD00041136
CAS Number: 4430-51-7
UN: 2923
ADR: 8,III