trans-3-Hexen-1-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: trans-Hex-3-en-1-ol

W435600-1KG-K W435600-100G-K
SIALW435600-1KG-KEA 988 EUR
SIALW435600-1KG-K SIALW435600-100G-K
trans-3-Hexen-1-ol, Sigma-Aldrich®
trans-3-Hexen-1-ol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 155…156 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.515 g/cm³ (20 °C)
MDL Number: MFCD00002960
CAS Number: 928-97-2
EINECS: 213-193-3
UN: 1987
ADR: 3,III
Merck Index: 12,04737

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR