N-(5-Bromopentyl)phthalimide, Sigma-Aldrich®
Supplier: Merck
722499-25G
722499-5G
SIAL722499-25GEA
171
EUR
SIAL722499-25G
SIAL722499-5G
N-(5-Bromopentyl)phthalimide, Sigma-Aldrich®
N-(5-Bromopentyl)phthalimide
Formula:
C₁₃H₁₄BrNO₂ MW: 296.16 g/mol |
MDL Number:
MFCD00060522 CAS Number: 954-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...