Thymine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
T0376-100G T0376-10G T0376-25G T0376-5G
SIALT0376-100GEA 426 EUR
SIALT0376-100G SIALT0376-10G SIALT0376-25G SIALT0376-5G
Thymine, Sigma-Aldrich®
Thymine
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Boiling Pt: 389 °C (1013 hPa)
Melting Pt: 218 °C
Density: 1.454 g/cm³ (25 °C)
MDL Number: MFCD00006026
CAS Number: 65-71-4
EINECS: 200-616-1
Merck Index: 12,09539