3-(Aminomethyl)pyridine
Supplier: Thermo Scientific
Synonyms: 3-Pyridylmethylamine, (3-Pyridyl)methylamine, 3-Picolamine, Pyridin-3-ylmethanamine, 3-Picolylamine
CC04113.10 CC04113.1
MAYBCC04113.10EA 0 EUR
MAYBCC04113.10 MAYBCC04113.1
3-(Aminomethyl)pyridine
3-(Aminomethyl)pyridine
Formula: C₆H₈N₂
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1,0712 g/cm³ (20 °C)
MDL Number: MFCD00006412
CAS Number: 3731-52-0
EINECS: 223-091-0