Cytosine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 6-Aminopyrimidin-2(1H)-one, 4-Amino-2-pyrimidinol
C3506-100G C3506-10G C3506-1G C3506-25G C3506-5G
SIALC3506-100GEA 998 EUR
SIALC3506-100G SIALC3506-10G SIALC3506-1G SIALC3506-25G SIALC3506-5G
Cytosine, Sigma-Aldrich®
Cytosine
Formula: C₄H₅N₃O
MW: 111.1 g/mol
Melting Pt: 313…316 °C
Density: 0.99771 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006034
CAS Number: 71-30-7
EINECS: 200-749-5
Merck Index: 13,02824