Uric acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione

U0881-100G U0881-10G U0881-25G
SIALU0881-100GEA 672 EUR
SIALU0881-100G SIALU0881-10G SIALU0881-25G
Uric acid, Sigma-Aldrich®
Uric acid
Formula: C₅H₄N₄O₃
MW: 168.11 g/mol
Melting Pt: 436 °C
Density: 1.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005712
CAS Number: 69-93-2
EINECS: 200-720-7
Merck Index: 12,10014

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR