2-Amino-6-ethylpyridine

Supplier: Thermo Scientific

Synonyms: 6-Ethylpyridin-2-amine, 6-Ethyl-2-pyridylamine

BTB11177.1 BTB11177.10 BTB11177.50
MAYBBTB11177.1EA 0 EUR
MAYBBTB11177.1 MAYBBTB11177.10 MAYBBTB11177.50
2-Amino-6-ethylpyridine
2-Amino-6-ethylpyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00218265
CAS Number: 21717-29-3
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR