2-Bromo-1,4-dimethoxybenzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Bromo-2,5-dimethoxybenzene
M-Clarity™ quality level = MQ100
252743-25G 252743-5G
SIAL252743-25GEA 57.6 EUR
SIAL252743-25G SIAL252743-5G
2-Bromo-1,4-dimethoxybenzene, Sigma-Aldrich®
2-Bromo-1,4-dimethoxybenzene
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 107 °C (0.2 mmHg)
Density: 1.445 g/cm³ (15 °C)
MDL Number: MFCD00008355
CAS Number: 25245-34-5
EINECS: 246-756-7