4-Chloro-2-nitrobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
C4753-250MG
C4753-5G
SIALC4753-250MGEA
59.5
EUR
SIALC4753-250MG
SIALC4753-5G
4-Chloro-2-nitrobenzaldehyde, Sigma-Aldrich®
4-Chloro-2-nitrobenzaldehyde
Formula:
C₇H₄ClNO₃ MW: 185.57 g/mol |
MDL Number:
MFCD00056104 CAS Number: 5551-11-1 EINECS: 226-915-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...