3-Chloro-4-methoxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Chloro-4-methoxybenzaldehyde
M-Clarity™ quality level = MQ100
565040-1G 565040-5G
SIAL565040-1GEA 71.1 EUR
SIAL565040-1G SIAL565040-5G
3-Chloro-4-methoxybenzaldehyde, Sigma-Aldrich®
3-Chloro-4-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00016601
CAS Number: 4903-09-7
EINECS: 225-532-2