Dimethylmalonic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,2-Dimethylmalonic acid
M-Clarity™ quality level = MQ200
D168009-25G D168009-5G
SIALD168009-25GEA 85.8 EUR
SIALD168009-25G SIALD168009-5G
Dimethylmalonic acid, Sigma-Aldrich®
Dimethylmalonic acid
Formula: C₅H₈O₄
MW: 132.12 g/mol
MDL Number: MFCD00004193
CAS Number: 595-46-0
EINECS: 209-867-1