Palladium(II) sulphate ≥98%, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
205788-1G
205788-5G
SIAL205788-1GEA
188
EUR
SIAL205788-1G
SIAL205788-5G
Palladium(II) sulphate ≥98%, Sigma-Aldrich®
Palladium (II) sulphate
Formula:
O₄PdS MW: 202.48 g/mol |
MDL Number:
MFCD00011173 CAS Number: 13566-03-5 EINECS: 236-957-8 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...