L-Threonine, Sigma-Aldrich®

Supplier: Merck

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

89179-10G 89179-250G 89179-50G
SIAL89179-10GEA 61.3 EUR
SIAL89179-10G SIAL89179-250G SIAL89179-50G
L-Threonine, Sigma-Aldrich®
L-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR