1,2,3,4-Tetrahydroquinoline, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

87350-100ML 87350-500ML
SIAL87350-100MLEA 59.4 EUR
SIAL87350-100ML SIAL87350-500ML
1,2,3,4-Tetrahydroquinoline, Sigma-Aldrich®
1,2,3,4-Tetrahydroquinoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00006693
CAS Number: 635-46-1
EINECS: 211-237-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR