Ethyl-4-chlorobutyrate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A11070.22
A11070.36
A11070.22EA
46.6
EUR
A11070.22
A11070.36
Ethyl-4-chlorobutyrate ≥98%
Ethyl 4-chlorobutyrate
Purity: 98%
CAS No.: 3153-36-4
Molecular Formula: Cl(CH2)3CO2CH2CH3
Formula Weight: 150.61
Flash Point: 76°(169°F)
Refractive Index: 1.4320
Boiling Point: 185-186°
Beilstein Reference: 1098810
MDL No.: MFCD00001004
Formula:
C₆H₁₁ClO₂ Boiling Pt: 185…186 °C Density: 1.077 Flash Pt: 76 °C (169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001004 CAS Number: 3153-36-4 EINECS: 221-591-3 |