2-Amino-4,6-difluorobenzothiazole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4,6-Difluorobenzothiazol-2-ylamine

M-Clarity™ quality level = MQ100

683337-1G 683337-5G
SIAL683337-1GEA 32.6 EUR
SIAL683337-1G SIAL683337-5G
2-Amino-4,6-difluorobenzothiazole, Sigma-Aldrich®
2-Amino-4,6-difluorobenzothiazole
Formula: C₇H₄F₂N₂S
MW: 186.18 g/mol
MDL Number: MFCD00276099
CAS Number: 119256-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR