Isoamyl butyrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Isopentyl butyrate, Isoamyl Butyrate

M-Clarity™ quality level = MQ100

W206016-1KG-K W206016-20KG-K W206016-4KG-K
SIALW206016-1KG-KEA 236 EUR
SIALW206016-1KG-K SIALW206016-20KG-K SIALW206016-4KG-K
Isoamyl butyrate, Sigma-Aldrich®
Isoamyl butyrate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
MDL Number: MFCD00044888
CAS Number: 106-27-4
EINECS: 203-380-8
UN: 2620
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR