1-Bromo-3,5-difluoro-2-iodobenzene

Supplier: Thermo Scientific

TL00647.1 TL00647.10 TL00647.50
MAYBTL00647.1EA 0 EUR
MAYBTL00647.1 MAYBTL00647.10 MAYBTL00647.50
1-Bromo-3,5-difluoro-2-iodobenzene
1-Bromo-3,5-difluoro-2-iodobenzene
Formula: C₆H₂BrF₂I
MW: 318.89 g/mol
MDL Number: MFCD00042183
CAS Number: 175278-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR