1-Ethyl-3-methylimidazolium acetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Ethyl-3-methylimidazolium Acetate

M-Clarity™ quality level = MQ100

689483-50G 689483-5G
SIAL689483-50GEA 811 EUR
SIAL689483-50G SIAL689483-5G
1-Ethyl-3-methylimidazolium acetate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium acetate
Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
MDL Number: MFCD06798186
CAS Number: 143314-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR