1-Ethyl-3-methylimidazolium dimethylphosphate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
671657-50G 671657-5G
SIAL671657-50GEA 252 EUR
SIAL671657-50G SIAL671657-5G
1-Ethyl-3-methylimidazolium dimethylphosphate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium dimethylphosphate
Formula: C₈H₁₇N₂O₄P
MW: 236.21 g/mol
CAS Number: 945611-27-8