3-Cyclopentyloxy-4-methoxyaniline
Supplier: Thermo Scientific
KM09285.250
KM09285.1
MAYBKM09285.250EA
0
EUR
MAYBKM09285.250
MAYBKM09285.1
3-Cyclopentyloxy-4-methoxyaniline
3-Cyclopentyloxy-4-methoxyaniline
Formula:
C₁₂H₁₇NO₂ |
CAS Number:
154464-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...