Trimethylsilyl isocyanate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
252646-25G 252646-5G
SIAL252646-25GEA 243 EUR
SIAL252646-25G SIAL252646-5G
Trimethylsilyl isocyanate, Sigma-Aldrich®
Trimethylsilyl isocyanate
Formula: C₄H₉NOSi
MW: 115.21 g/mol
MDL Number: MFCD00001993
CAS Number: 1118-02-1
EINECS: 214-256-8