1-Benzyl-4-piperidone, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone
M-Clarity™ quality level = MQ200
B29806-100G B29806-25G
SIALB29806-100GEA 75.6 EUR
SIALB29806-100G SIALB29806-25G
1-Benzyl-4-piperidone, Sigma-Aldrich®
1-Benzyl-4-piperidone
Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
MDL Number: MFCD00006192
CAS Number: 3612-20-2
EINECS: 222-782-4