1-Butyl-3-methylimidazolium hydrogen sulphate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium hydrogensulphate
57457-1KG-F 57457-100G-F
SIAL57457-1KG-FEA 1950 EUR
SIAL57457-1KG-F SIAL57457-100G-F
1-Butyl-3-methylimidazolium hydrogen sulphate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium hydrogen sulphate
Formula: C₈H₁₆N₂O₄S
MW: 236.29 g/mol
MDL Number: MFCD06798197
CAS Number: 262297-13-2
UN: 3265
ADR: 8,III