(±)-2-Bromoisovaleric acid

Supplier: Thermo Scientific

Synonyms: 2-Bromo-3-methylbutyricacid, 2-Bromo-3-methylbutyric acid

RDP00014.1 RDP00014.10 RDP00014.50
MAYBRDP00014.1EA 0 EUR
MAYBRDP00014.1 MAYBRDP00014.10 MAYBRDP00014.50
(±)-2-Bromoisovaleric acid
(±)-2-Bromoisovaleric acid
Formula: C₅H₉BrO₂
MW: 181.03 g/mol
MDL Number: MFCD00004210
CAS Number: 565-74-2
EINECS: 209-291-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR