Phenol ≥99%, crystals ACS
Supplier: VWR Chemicals
0735-500GEA 184 EUR
0735-500G 0735-250G 0735-1KG
Phenol ≥99%, crystals ACS
Phenol
Formula: C₆H₅OH
MW: 94,11 g/mol
Boiling Pt: 181,8 °C (1013 hPa)
Melting Pt: 40,8 °C
Density: 1,06 g/cm³ (20 °C)
Flash Pt: 81 °C (closed cup)
MDL Number: MFCD00002143
UN: 1671
ADR: 6.1,II
Order Now
Specification Test Results
Clarity of Solution Pass
Freezing Range ≥40.5 °C
Moisture (KF) ≤0.5%
Purity ≥99%
Residue After Evaporation ≤0.05%