Diethyl bromomalonate ≥90%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Diethyl 2-bromomalonate

A10965.14 A10965.18 A10965.30
A10965.14EA 31.6 EUR
A10965.14 A10965.18 A10965.30
Diethyl bromomalonate ≥90%
Diethyl bromomalonate

Purity: 90+% CAS No.: 685-87-0 Molecular Formula: BrCH(CO2CH2CH3)2 Formula Weight: 239.07 Flash Point: 111°(231°F) Refractive Index: 1.4550 Boiling Point: 233-235° Beilstein Reference: 509768 UN No.: 3265 MDL No.: MFCD00009138

Formula: C₇H₁₁BrO₄
MW: 239.07 g/mol
Boiling Pt: 233…235 °C
Density: 1.402
Flash Pt: 111 °C (231 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009138
CAS Number: 685-87-0
EINECS: 211-683-1
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR