Diethyl bromomalonate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Diethyl 2-bromomalonate

M-Clarity™ quality level = MQ100

D91209-100G D91209-25G
SIALD91209-100GEA 270 EUR
SIALD91209-100G SIALD91209-25G
Diethyl bromomalonate, Sigma-Aldrich®
Diethyl bromomalonate
Formula: C₇H₁₁BrO₄
MW: 239.07 g/mol
MDL Number: MFCD00009138
CAS Number: 685-87-0
EINECS: 211-683-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR