Dimethyl chloromalonate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Chloromalonic acid dimethyl ester, Dimethyl 2-chloromalonate

M-Clarity™ quality level = MQ200

374539-100G 374539-25G
SIAL374539-100GEA 535 EUR
SIAL374539-100G SIAL374539-25G
Dimethyl chloromalonate, Sigma-Aldrich®
Dimethyl chloromalonate
Formula: C₅H₇ClO₄
MW: 166.56 g/mol
MDL Number: MFCD00042837
CAS Number: 28868-76-0
EINECS: 249-275-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR