(S)-(-)-1-(1-Naphthyl)ethylamine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(S)-(-)-alpha-(1-Naphthyl)ethylamine
M-Clarity™ quality level = MQ100
237450-1G
237450-5G
SIAL237450-1GEA
57.6
EUR
SIAL237450-1G
SIAL237450-5G
(S)-(-)-1-(1-Naphthyl)ethylamine, Sigma-Aldrich®
(S)-(-)-1-(1-Naphthyl)ethylamine
Formula:
C₁₂H₁₃N MW: 171.24 g/mol |
MDL Number:
MFCD00064179 CAS Number: 10420-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...