3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide
M-Clarity™ quality level = MQ200
M5655-1G M5655-100MG M5655-500MG
SIALM5655-1GEA 155 EUR
SIALM5655-1G SIALM5655-100MG SIALM5655-500MG
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), Sigma-Aldrich®
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formula: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5