Diethyl cyclopropane-1,1-dicarboxylate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Diethyl 1,1-cyclopropanedicarboxylate, Diethyl 1,1-cyclopropane dicarboxyliate
M-Clarity™ quality level = MQ200
147710-25G 147710-5G
SIAL147710-25GEA 317 EUR
SIAL147710-25G SIAL147710-5G
Diethyl cyclopropane-1,1-dicarboxylate, Sigma-Aldrich®
Diethyl cyclopropane-1,1-dicarboxylate
Formula: C₉H₁₄O₄
MW: 186.21 g/mol
MDL Number: MFCD00001283
CAS Number: 1559-02-0
EINECS: 216-321-6