Diphenyl sulphoxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Phenyl sulphoxide, sulphinyldibenzene

M-Clarity™ quality level = MQ100

P35405-100G P35405-25G
SIALP35405-100GEA 93.7 EUR
SIALP35405-100G SIALP35405-25G
Diphenyl sulphoxide, Sigma-Aldrich®
Diphenyl sulphoxide
Formula: C₁₂H₁₀OS
MW: 202.28 g/mol
MDL Number: MFCD00002085
CAS Number: 945-51-7
EINECS: 213-415-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR