3-Aminophenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific
Warning
Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester
367590010 367590050
ACRO367590010EA 69.6 EUR
ACRO367590010 ACRO367590050
3-Aminophenylboronic acid pinacol ester 97%
3-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03453668
CAS Number: 210907-84-9
Order Now
Specification Test Results
Appearance Light brown Powder or crystals
Infrared spectrum Conforms
Melting point 89 to 94 °C
GC ≥96.0 %